Renilla luciferase assay system
Supplier: Promega
Certificates
About this item
Renilla Luciferase assay system is designed to provide a fast, sensitive method of detecting luciferase from sea pansy (Renilla reniformis). The system is a convenient alternative to firefly (Photinus pyralis) reporter systems and is designed to yield reliable, linear results for a concentration range over 7 orders of magnitude.
- Linear range extends over 7 logs
- Low background provides increased sensitivity
- Detect 10⁻¹⁹ moles of Renilla luciferase
The Renilla Luciferase Assay System is formulated with a proprietary composition that significantly reduces the effect of coelenterazine autoluminescence compared to other reagents, making the reagent orders of magnitude more sensitive than published methods. It enables measurements with wildtype and synthetic hRluc genes for primary expression or internal normalisation measurements of gene expression.
Delivery: System includes Renilla luciferase assay substrate, Renilla luciferase assay buffer and Renilla luciferase assay lysis buffer (5X).
Frequently Bought Together