Order Entry
Portugal
Orders LinkContactUsLinkComponent
PCR markers

PCR markers

PCR markers, 250 µl
Catalog # PROMG31610250
Supplier:   Promega
PCR markers
Catalog # PROMG31610250
Supplier:  Promega
CAS Number:  
PCR markers
PCR markers

PCR markers, 250 µl

CAS Number:  
Price
148,00 €
Log in for availability
1 * 250 µl
Product Details & Documents

PCR markers have six bands of equal intensity of 50, 150, 300, 500, 750 and 1000 bp.

  • Six equal intensity bands
  • Load 5 μl/lane

The BenchTop PCR marker may be run on polyacrylamide gels with less loading volume. However, additional bands may be visible compared to those visible on agarose gels.

Storage buffer: 10 mM Tris-HCl (pH 8.0), 1 mM EDTA.

Delivery information: Supplied with 6X blue/orange loading dye at a concentration of ~0,06 µg/µl.

Caution: Not intended for use in quantitative analysis.

Specifications

Specifications

  • Pk:
    250 µl
  • Description:
    PCR markers

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.