RNA purification, SV 96 Total RNA isolation system
Supplier: Promega
Certificates
About this item
SV 96 Total RNA Isolation System provides a high-throughput technique to prepare intact RNA from tissue and cultured cells. Total RNA can be purified from 96 samples in less than an hour without centrifugation.
- Vacuum-based method - no centrifugation required
- Plate design prevents cross-contamination during sample processing
- Designed for both manual and automated RNA purification
SV Total RNA Isolation System incorporates a DNase treatment step directly on the mini column membrane. This step substantially reduces genomic DNA contamination, which can interfere with amplification-based methodologies. Purification is achieved without the use of phenol:chloroform extractions or ethanol precipitations, and there is no DNase carry over in the final RNA preparation.
Protocols are available for Beckman Coulter and PerkinElmer instruments.
Delivery: Kit includes binding plates; elution plates; plate sealers; RNA lysis buffer (RLA); β-mercaptoethanol (97,4%); DNase I (lyophilised); MnCl2 (0,09 M); yellow core buffer; DNase stop solution (DSA) (concentrated); RNA wash solution (RWA); nuclease-free water.
Caution: The SV 96 Total RNA Isolation System requires a vacuum pump capable of generating 15–20 inches of Hg or equivalent.
Frequently Bought Together