Genomic DNA purification kits, MagneSil® Genomic, Fixed-Tissue system
PROMMD1490
: Promega
:
- Pk:100 Tests
- Description:MagneSil® Genomic, Fixed-Tissue System
The MagneSil® Genomic, Fixed-Tissue System provides a fast, simple technique for extracting genomic DNA from formalin-fixed, paraffin-embedded (FFPE) tissue. After an overnight Proteinase K digestion, genomic DNA can be manually purified from FFPE thin tissue sections in less than an hour, and up to 12 samples can be processed at a time using the MagneSphere® technology magnetic separation stand (12-position).
- No centrifugation of lysate required
- Amplifiable DNA from 10 µM FFPE tissue slices
- Targets as large as 450 to 1800 bp can be amplified
- Multiplex PCR can be performed using DNA purified with this system
: Kit includes Proteinase K, DTT, Incubation Buffer, Elution Buffer, 2X Wash Buffer, Lysis Buffer and resin.