DNA polymerase I large (Klenow) fragment mini kit
Catalog # PROMU13000150
Supplier: Promega
CAS Number:
Specifications
- Pk:150 EU
- Description:DNA Polymerase I Large (Klenow) Fragment Mini Kit
Specifications
About this item
The DNA polymerase I large (Klenow) fragment mini kit provides polymerase and dNTPs ready to use in a variety of applications.
Delivery: Kit includes polymerase I large (Klenow) fragment (150 U), DNA polymerase 10X buffer (1 ml), 5 µmol (1×500 µl) each of dATP, dGTP, dTTP and dCTP (10 mM in water).