Ribonucleotide solution, Ribo m⁷G Cap Analog
Supplier: Promega
Certificates
About this item
The Ribo m⁷G Cap Analog is a modified ribonucleotide with the structure (m⁷G(5´)ppp(5´)G). This methylated ribonucleotide can be incorporated onto the 5´-end of transcripts synthesised in vitro and simulates the 7-methyl guanosine 5´-cap structure found on most eukaryotic mRNA molecules.
- Enhances translation efficiency in reticulocyte-based reactions
- Protects RNA from intracellular digestion
- Can be used with Riboprobe® or RiboMAX™ RNA production systems
- Supplied at a concentration of 40 mM
Frequently Bought Together