Order Entry
Portugal
Orders LinkContactUsLinkComponent
Buffer pack, MULTI-CORE™

Buffer pack, MULTI-CORE™

MULTI-CORE™ buffer pack, 3×1 ml (10X), 3 mL
Catalog # PROMR99913X1
Supplier:   Promega
Buffer pack, MULTI-CORE™
Catalog # PROMR99913X1
Supplier:  Promega
CAS Number:  
Buffer pack, MULTI-CORE™
Buffer pack, MULTI-CORE™

MULTI-CORE™ buffer pack, 3×1 ml (10X), 3 mL

CAS Number:  
Price
57,00 €
Log in for availability
1 * 3 mL
Product Details & Documents

The MULTI-CORE™ buffer pack contains convenient aliquots of the universal restriction enzyme 10X buffer. It is formulated to provide simple buffering conditions for performing multiple digestions.

  • Enables digestion of DNA with more than one enzyme in a single reaction
  • Only modest adjustments in the amount of enzyme used will ensure complete multiple digestions
  • Storage conditions: −30 to −10 °C

1X MULTI-CORE™ buffer concentrated solution of 25 mM Tris-acetate (pH 7,5), 100 mM potassium acetate, 100 mM magnesium acetate, 1 mM DTT.

Delivery information: Pack includes 3×1 ml MULTI-CORE™ 10X buffer, 10 mg/ml acetylated BSA.

Specifications

Specifications

  • Pk:
    3 mL
  • Description:
    MULTI-CORE™ buffer pack, 3×1 ml (10X)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.