Order Entry
Portugal
Orders LinkContactUsLinkComponent
Buffer pack, 4-CORE®

Buffer pack, 4-CORE®

4-CORE® buffer pack, includes buffers A, B, C and D (10X), 4 mL
Catalog # PROMR992104X1
Supplier:   Promega
Buffer pack, 4-CORE®
Catalog # PROMR992104X1
Supplier:  Promega
CAS Number:  
Buffer pack, 4-CORE®
Buffer pack, 4-CORE®

4-CORE® buffer pack, includes buffers A, B, C and D (10X), 4 mL

CAS Number:  
Price
69,70 €
Log in for availability
1 * 4 mL
Product Details & Documents

The 4-CORE® buffer pack contains convenient aliquots of restriction enzyme 10X Buffers A, B, C and D. The majority of restriction enzymes have optimal activity in one of these four 10X reaction buffers.

  • Storage conditions: −30 to −10 °C

Note: All restriction enzymes are supplied with 10 mg/ml acetylated BSA. Although BSA is not absolutely required for activity, it has been shown to enhance activity of many restriction enzymes. Adding BSA to all restriction digests at a final concentration of 0,1 mg/ml.

1X MULTI-CORE™ buffer concentrated solution of 25 mM Tris-acetate (pH 7,5), 100 mM potassium acetate, 100 mM magnesium acetate, 1 mM DTT.

Delivery information: Pack includes 10X (A, B, C, D) buffer (1 ml), 10 mg/ml acetylated BSA.

Specifications

Specifications

  • Pk:
    4 mL
  • Description:
    4-CORE® buffer pack, includes buffers A, B, C and D (10X)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.