Order Entry
Portugal
ContactUsLinkComponent
Test kits, photometric, Bitter units, LCK cuvette tests
Test kits, photometric, Bitter units, LCK cuvette tests
Catalog # HACHLCK241
Supplier:  Hach
CAS Number:  
undefined
Test kits, photometric, Bitter units, LCK cuvette tests
Catalog # HACHLCK241
Supplier:  Hach
CAS Number:  

Specifications

  • Chemical method:
    Analogous MEBAK-Method
  • Pk:
    25 Tests
  • Description:
    Bitter units
  • Seq:
    205
  • Type:
    LCK241
  • For unit:
    DR5000, DR6000
  • Range:
    ≥ 2 Bitter units

Specifications

About this item

For use with your Hach DR6000 spectrophotometer.

  • Ready-to-use, high-precision pre-dosed cuvette tests
  • Reduction of errors due to bar-coded cuvettes
  • RFID technology to document the shelf life of the cuvettes and ensure traceability of your analyses
  • No reagent blank needed
  • Innovative cuvette and reagent technique

LCK241 offers you the ability to measure international Bitter Units with less hassle and no chemical waste. Additionally, these cuvettes provide high quality results free from potential external influencers. They are designed to simplify your brewing analysis, with accurate results, every time.

Storage conditions: 15 to 25 °C

Certifications: MEBAK II