Restriction enzymes, PstI
Supplier: Promega
Certificates
About this item
These restriction enzymes are capable of digesting DNA in 15 minutes or less. Recognises C TGCA˅G and G˄ACGT C.
- Active and capable of digestion directly in GoTaq® Green Master Mix
- Blue or white cloning qualified, provides a higher level of quality control
- With incubation at 37 °C in buffer H
- Source: Recombinant E. coli strain
Storage Buffer: 10 mM Tris-HCl (pH 7,3), 300 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,15% Triton® X-100, 0,5 mg/ml BSA, 50% glycerol.
Delivery: The kit includes 1×1 ml buffer H 10X buffer, 1×150 µl bovine serum albumin, acetylated with concentration of 10 mg/ml, 1×3,000 units PstI and 1×0,25 ml MULTI-CORE™ 10X buffer.
Specifications
- Source:E. coli
- Storage conditions:−20 °C
- Enzyme name:PstI
- Buffer supplied with enzyme:Buffer H
- Buffer activity:% Activity (37°C) in 4 CORE® Buffers A: 10…25% – B: 50…75% – C: 50…75% – D: 50…75%; MULTI-CORE™ Buffer 25…50%
- Enzyme assay temperature:37°C
Specifications
Frequently Bought Together