Order Entry
Portugal
ContactUsLinkComponent
Anti-phospho-SLP76 Tyr145 Rabbit Polyclonal Antibody
Anti-phospho-SLP76 Tyr145 Rabbit Polyclonal Antibody
Catalog # BIRBORB158424-100
Supplier:  Biorbyt
Anti-phospho-SLP76 Tyr145 Rabbit Polyclonal Antibody
Catalog # BIRBORB158424-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    phospho-SLP76 Tyr145
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Storage temperature:
    Store at 4 °C for one month and at –20 °C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    KLH conjugated synthetic peptide derived from human phospho-SLP76 (Tyr145)
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: phospho-SLP76 Tyr145
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human