Anti-BHLHB5 Rabbit Polyclonal Antibody
Catalog # BIRBORB155838-200
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:BHLHB5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Storage temperature:Store at 4 °C for one month and at –20 °C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:KLH conjugated synthetic peptide derived from human BHLHB5
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: BHLHB5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat