High yield wheat germ protein expression system, TNT® SP6
PROML3260
: Promega
:
- Pk:40 Tests
- Description:TNT® SP6 high yield wheat germ protein expression system
- Storage temperature:–70 °C
The TNT® SP6 high yield wheat germ protein expression system contains optimised wheat germ extract in a single-tube, coupled transcription/translation system that allows you to express proteins in only two hours from plasmid or PCR-generated templates.
- Express soluble, full length protein from plasmid or PCR-generated templates quickly
- Generate 10 to 20 fold more protein than with other cell-free systems
Protein is synthesised in concentrations in the range of 10 to 100 μg/ml and can be used in multiple applications, including high throughput analysis.
: Includes 4×300 µl TNT® SP6 high yield wheat germ master mix and 1×1250 µl nuclease-free water.