Restriction enzymes, EcoRI
PROMR601715KU
: Promega
:
- Concentration:12 U/µl
- Pk:15.000 EU
- Source:E. coli RY 13
- Description:EcoRI
- Seq:02
- Enzyme name:EcoRI
- Buffer supplied with enzyme:Buffer H
- Buffer activity:% Activity (37°C) in 4 CORE® Buffers A: 25…50% – B: 50…75% – C: 50…75% – D: 50…75%; MULTI-CORE™ Buffer 100%**
- Enzyme assay temperature:37°C
These EcoRI restriction enzyme recognises and cut G˅AATT C and C TTAA^G at 37 °C in its own unique buffer.
- Capable of digesting DNA in 15 minutes or less
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Source: Escherichia coli RY 13
Storage buffer: 10 mM Tris-HCl (pH 7.4), 400 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,15% Triton® X-100, 0,5 mg/ml BSA, 50% glycerol.
: Kit includes 1×1 ml buffer H 10X buffer, 1×750 µl bovine serum albumin (acetylated) and 1×5,000 units EcoRI.