Order Entry
Portugal
ContactUsLinkComponent
Cell viability assay, CellTiter-Glo® One Solution Assay
 :  Promega
undefined
Cell viability assay, CellTiter-Glo® One Solution Assay
 :  Promega
  • Storage conditions:
    Store the CellTiter-Glo One Solution Assay below –10 °C. CellTiter-Glo One Solution Assay can be stored at 4°C for 48 hours or at 22 °C for 10 hours with ~10 - 12% loss of activity. CellTiter-Glo One Solution Assay can withstand two additional freeze-thaw cycles after the first thaw with approximately 10% loss of activity with each additional freeze-thaw cycle.
  • Storage temperature:
    –30...–10 °C
Frequently Bought Together