Apoptosis assays, Caspase-Glo® 9 assay
Supplier: Promega
Certificates
About this item
The Caspase-Glo® 9 assay is a homogeneous luminescent assay that measures caspase-9 activities. The assay provides a proluminescent caspase-9 substrate in a buffer system optimised for caspase-9 activity, luciferase activity and cell lysis. The addition of a single Caspase-Glo® 9 reagent results in cell lysis, followed by caspase cleavage of the substrate and generation of 'glow-type' luminescent signal. The signal is proportional to caspase-9 activity.
- Add-mix-measure protocol amenable to automation
- Low background luminescence results in excellent signal-to-noise ratios, allows assay to be performed in 96- or 384-well formats
- No sample preparation, manipulation and extended incubation times required, maximum sensitivity can be achieved in as little as 30 minutes to 1 hour
- Delivers excellent Z´ factor values in cell and purified enzyme models
- Could multiplex with other cell-based assays
Delivery: The complete assay kit includes Caspase-Glo® 9 buffer, Caspase-Glo® 9 substrate and MG-132 inhibitor.
Frequently Bought Together