Coupled Reticulocyte Lysate Systems, TnT®
Supplier: Promega
Certificates
About this item
TnT® Coupled Reticulocyte Lysate System offers researchers an alternative for eukaryotic cell-free protein expression. It greatly simplifies the process and reduces the time required to obtain in vitro translation results.
- Study protein:protein, protein:nucleic acid and protein expression interactions
- Generate proteins in one hour versus days
Delivery: TnT® Rabbit Reticulocyte Lysate is supplied in 200 µl aliquots. Each system contains sufficient reagents to perform
approximately 40×50 µl translation reactions. Includes:
1 ml TnT® Rabbit Reticulocyte Lysate
90 µl TnT® Reaction Buffer
60 µl TnT® T3, T7 or SP6 RNA Polymerase (2×30 µl in dual systems)
5 µg Luciferase T3, T7 or SP6 Control DNA, 0,5 mg/ml
50 µl Amino Acid Mixture, minus Methionine, 1 mM
50 µl Amino Acid Mixture, minus Leucine, 1 mM
50 µl Amino Acid Mixture, minus Cysteine, 1 mM
250 µl Luciferase Assay Reagent
1× Luciferase Assay Wells (set of 3)
100 µl Magnesium Acetate (only with Cat. Nos. L4600 and L4601)
approximately 40×50 µl translation reactions. Includes:
1 ml TnT® Rabbit Reticulocyte Lysate
90 µl TnT® Reaction Buffer
60 µl TnT® T3, T7 or SP6 RNA Polymerase (2×30 µl in dual systems)
5 µg Luciferase T3, T7 or SP6 Control DNA, 0,5 mg/ml
50 µl Amino Acid Mixture, minus Methionine, 1 mM
50 µl Amino Acid Mixture, minus Leucine, 1 mM
50 µl Amino Acid Mixture, minus Cysteine, 1 mM
250 µl Luciferase Assay Reagent
1× Luciferase Assay Wells (set of 3)
100 µl Magnesium Acetate (only with Cat. Nos. L4600 and L4601)
Specifications
- Storage temperature:–70 °C (exception: luciferase assay wells should be stored at room temperature)