T7 quick protein expression system, for PCR, TNT®
Catalog # PROML5540
Supplier: Promega
CAS Number:
Specifications
- Pk:40 Tests
- Description:TnT® T7 Quick for PCR
- Storage temperature:–70 °C
Specifications
About this item
The TnT® T7 Quick for PCR is a simple, single-tube coupled transcription/translation system designed for optimum protein expression directly from PCR templates.
- Express protein directly from PCR, no clean up needed
- Up to five times more protein than standard translation reactions with linear templates
- Includes recombinant RNasin® ribonuclease inhibitor
The PCR-generated DNA can be used directly from the amplification reaction or purified by numerous commercially available kits and traditional methods. For most PCR templates, the reactions produce up to 5 times more protein than other commercially available kits.
Delivery: Includes methionine, TnT® T7 PCR quick master mix , nuclease-free water.