Order Entry
Portugal
ContactUsLinkComponent
RealTime-Glo™ MT Cell viability assay
RealTime-Glo™ MT Cell viability assay
Supplier:  Promega
Certificates
undefined
RealTime-Glo™ MT Cell viability assay
Supplier:  Promega
Specifications
  • Storage conditions:
    Store protected from light. Avoid prolonged exposure to light. Avoid multiple freeze-thaw cycles.
  • Storage temperature:
    −20 °C