Cell lysis reagent, FastBreak™
Supplier: Promega
Certificates
About this item
FastBreak™ cell lysis reagent is provided as a 10X concentrate and contains a proprietary non ionic detergent to facilitate lysis. Recombinant proteins can be directly screened in the cell extract or purified by the addition of the appropriate affinity matrix such as the MagneHis™ protein purification system.
- Eliminate centrifugation and mechanical disruption
- Simple, add-and-incubate protocol
- Compatible with manual or robotic platforms
Frequently Bought Together