CYP3A4 (luciferin-PPXE) DMSO tolerant assay and screening systems, P450-Glo™
Supplier: Promega
Certificates
About this item
The P450-Glo CYP3A4 with luciferin-PPXE assays provide a homogeneous, luminescent method for measuring cytochrome CYP3A4 activity in hepatocytes.
- Broad dynamic range, low background and better sensitivity for less ambiguous data
- Luminescent output eliminates interference from fluorescent test compounds
- Optimal data quality for microplate reader assay
- Simplified, homogeneous experimental protocol
The P450-Glo CYP3A4 with luciferin-PPXE screening system provides a complete set of reagents for performing luminescent cytochrome P450 assays. The membranes are prepared from baculovirus-infected insect cells and contain human cytochrome P450, P450 reductase, and cytochrome b5. It also contains a membrane fraction devoid of cytochrome P450 activity as a negative control. The assays are ideal for testing the effects of drugs and new chemical entities on cytochrome P450 enzyme activities.
Delivery: Assay system contains Luciferin-PPXE, 50 mM (1×15 or 2×15 µl); luciferin detection reagent (lyophilised) (1 vial); reconstitution buffer (1×10 or 1×50 ml). Screening system contains P450-Glo CYP3A4 assay includes Luciferin-PPXE, 50 mM (2×15 µl); luciferin detection reagent (lyophilised) (1 vial); reconstitution buffer (50 ml) and human CYP3A4 enzyme system includes CYP3A4 (1 pmol/µl), reductase and b5 (500 µl); control membranes (100 µl); potassium phosphate buffer, 1M (pH 7.4) (2×5 ml); luciferin-free water (50 ml); NADPH regeneration system solution A (2,75 ml); NADPH regeneration system solution B (0,6 ml).
Frequently Bought Together