CYP1A1 assay systems, P450-Glo™
: Promega
The P450-Glo CYP1B1 assays provide a homogeneous, luminescent method for measuring cytochrome CYP1B1 activity. These assays can be used to selectively measure CYP1A1 activity in hepatocytes where cross-reacting CYP1B1 and CYP3A7 are absent.
- Broad dynamic range, low background and better sensitivity for less ambiguous data
- Luminescent output eliminates interference from fluorescent test compounds
- Optimal data quality for microplate reader assay
- Simplified, homogeneous experimental protocol
: Assay system contains Luciferin-CEE, 5 mM (1×70 or 1×350 µl); luciferin detection reagent (lyophilised) (1 vial); reconstitution buffer (1×10 or 1×50 ml).
Frequently Bought Together