Order Entry
Portugal
ContactUsLinkComponent
Anti-CD3E Mouse Monoclonal Antibody [clone: PC3/188A]
Anti-CD3E Mouse Monoclonal Antibody [clone: PC3/188A]
Catalog # ABNOMAB14699
Supplier:  Abnova
undefined
Anti-CD3E Mouse Monoclonal Antibody [clone: PC3/188A]
Catalog # ABNOMAB14699
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD3e molecule, epsilon
  • Antigen symbol:
    CD3E
  • Clonality:
    Monoclonal
  • Clone:
    PC3/188A
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Epitope:
    152-168 aa
  • Form:
    Liquid
  • Gene ID:
    916
  • Storage buffer:
    In 10 mM PBS (0.05% BSA and 0.05% azide).
  • Storage temperature:
    Store at 4°C.
  • Immunogen:
    A synthetic peptide corresponding to amino acids 156-168 of the cytoplasmic domain of human CD3E.
  • Purification:
    Protein A purification
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against synthetic peptide of human CD3E.

Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: PC3/188A
Conjugation:
Epitope: 152-168 aa
Host: Mouse
Isotype: IgG1
Reactivity: Human