Order Entry
Portugal
Orders LinkContactUsLinkComponent
 

Restriction enzymes, Mbo I

Mbo I, 8 - 12 U/µl, 200 EU
Catalog # PROMR67110200
Supplier:   Promega
Restriction enzymes, Mbo I
Catalog # PROMR67110200
Supplier:  Promega
CAS Number:  
Restriction enzymes, Mbo I

Mbo I, 8 - 12 U/µl, 200 EU

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 200 UE
Product Details & Documents

These Mbo I restriction enzyme recognises ˅GATC and CTAG^ sites and cuts best at 37 °C in its own unique buffer.

  • Sensitive to dam methylation
  • DpnI is an unusual isoschizomer of MboI that will cut only if the recognition site is methylated by the dam methylation system
  • Sau3AI is an isoschizomer of MboI that is not sensitive to dam methylation
  • Source: Moraxella bovis ATCC 10900

Storage buffer: 10 mM Tris-HCl (pH 7,4), 50 mM KCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA and 50% glycerol.

 : Kit includes (1×200 U) Mbo I, (1×1 ml) buffer C 10X, (1×150 µl) bovine serum albumin (acetylated).

Specifications

Specifications

  • Concentration:
    8 - 12 U/µl
  • Pk:
    200 EU
  • Description:
    Mbo I
  • Cat. No.:
    PROMR67110200
  • Source:
    Moraxella bovis ATCC 10900
  • Enzyme name:
    Mbo I

Specifications