Order Entry
Portugal
ContactUsLinkComponent
 
Anti-TREM1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-10306R-A555
Supplier:  Bioss
Anti-TREM1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-10306R-A555
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Triggering Receptor Expressed on Myeloid Cells 1
  • Antigen symbol:
    TREM1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 555
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    54210
  • Antigen synonyms:
    Trem1|CD354|TREM-1|Triggering receptor expressed on myeloid cells 1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    150-200/234
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Stimulates neutrophil and monocyte-mediated inflammatory responses. Triggers release of pro-inflammatory chemokines and cytokines, as well as increased surface expression of cell activation markers. Amplifier of inflammatory responses that are triggered by bacterial and fungal infections and is a crucial mediator of septic shock.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TREM1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human