Order Entry
Portugal
ContactUsLinkComponent
 
Anti-SMC1A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-3568R-FITC
 :  Bioss
Anti-SMC1A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-3568R-FITC
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Structural Maintenance Of Chromosomes 1A
  • Antigen symbol:
    SMC1A
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    8243
  • Antigen synonyms:
    OTTHUMP00000061876|SMC1|DXS423E|Sb1.8|Smcb|CDLS2|SB1.8|SMC1alpha|DKFZp686L19178|SMC1A|KIAA0178|MGC138332|SMC1L1|SMCB
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    150-200/1233
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Involved in chromosome cohesion during cell cycle and in DNA repair. Central component of cohesin complex. The cohesin complex is required for the cohesion of sister chromatids after DNA replication. The cohesin complex apparently forms a large proteinaceous ring within which sister chromatids can be trapped. At anaphase, the complex is cleaved and dissociates from chromatin, allowing sister chromatids to segregate. The cohesin complex may also play a role in spindle pole assembly during mitosis. Involved in DNA repair via its interaction with BRCA1 and its related phosphorylation by ATM, or via its phosphorylation by ATR. Works as a downstream effector both in the ATM/NBS1 branch and in the ATR/MSH2 branch of S-phase checkpoint.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SMC1A
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat