SspI (Genome qualified)
About this item
These SspI restriction enzyme recognises AAT˅ATT and TTA^TAA sites and cuts best at 37 °C in its own unique buffer. Exhibit 100% activity in the recommended buffer and reaction conditions.
- Genome qualified to ensure optimal performance in genomic analysis applications
- Available at high concentration, containing 2 500 units of SspI at a concentration of 40 to 80 Units/µl
- Source: Sphaerotilus natans ATCC 13925
Storage buffer: 10 mM Tris-HCl (pH 7,4), 50 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA and 50% glycerol.
Delivery information: Kit includes 1×500 U SspI, 1×1 ml buffer E 10X, 1×0,25 ml MULTI-CORE™ 10X buffer, 1×150 µl bovine serum albumin (acetylated).
2 Options Available Below
Product Details & Documents
These SspI restriction enzyme recognises AAT˅ATT and TTA^TAA sites and cuts best at 37 °C in its own unique buffer. Exhibit 100% activity in the recommended buffer and reaction conditions.
- Genome qualified to ensure optimal performance in genomic analysis applications
- Available at high concentration, containing 2 500 units of SspI at a concentration of 40 to 80 Units/µl
- Source: Sphaerotilus natans ATCC 13925
Storage buffer: 10 mM Tris-HCl (pH 7,4), 50 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA and 50% glycerol.
Delivery information: Kit includes 1×500 U SspI, 1×1 ml buffer E 10X, 1×0,25 ml MULTI-CORE™ 10X buffer, 1×150 µl bovine serum albumin (acetylated).
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- PROMR66010500
- PROMR46042500
- Enzyme name:
- SspI
- SspI
- Source:
- Sphaerotilus natans ATCC 13925
- Sphaerotilus natans ATCC 13925
- Storage conditions:
- −20 °C
- −20 °C
Specifications
Product Family Options
Product Information
- ConcentrationPkAvailabilityPrice
- PROMR66010500SspISpecifications:
More
Specifications
Cat. No.PROMR66010500Enzyme nameSspISourceSphaerotilus natans ATCC 13925Storage conditions−20 °C - PROMR46042500SspISpecifications:
More
Specifications
Cat. No.PROMR46042500Enzyme nameSspISourceSphaerotilus natans ATCC 13925Storage conditions−20 °C
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



