Lambda DNA
Catalog # PROMD15010250
Supplier: Promega
CAS Number:
Specifications
- Pk:250 µG
- Description:Lambda DNA
- Environmentally Preferable:
Specifications
About this item
Restriction enzyme-digested lambda DNA may be used as a molecular weight size marker in gel analysis of nucleic acids. Lambda DNA is also a commonly used substrate in restriction enzyme activity assays and it also determined nucleotide sequence.