RT-PCR system, GoTaq® probe 2-step system
Catalog # A6110
Supplier: Promega
CAS Number:
Specifications
- Pk:200 Assays
- Application:RT-qPCR probe
- Description:GoTaq® probe 2-step RT-PCR system, 2 ml
Specifications
About this item
The GoTaq® 2-Step RT-qPCR system facilitates detection and relative quantification of RNA expression levels via a two-step RT-qPCR method using integrated components such as GoScript™ reverse transcription system and GoTaq® qPCR master mix. It includes an optimised reaction buffer and M-MLV reverse transcriptase designed to enable efficient synthesis of first-strand cDNA in preparation for PCR amplification. The cDNA product can be added directly to downstream qPCR amplification reactions.
- Sensitive detection for earlier quantitation of low and high copy number targets over a broad dynamic range
- Resistant to a wide range of PCR inhibitors
- Compatible with both fast and standard cycling methods on most Real-Time PCR instruments using TaqMan® and other probe assays
- Exceptional room temperature set-up is easy for automated and high throughput detection
Delivery: The complete system includes GoScript™ Reverse Transcriptase enzyme, GoScript™ reaction buffer(5X), GoTaq® qPCR master mix (2X), MgCl₂, PCR nucleotide mix, CXR reference dye, oligo(dT)15 primer, random primers, nuclease-free water and recombinant RNasin® ribonuclease inhibitor.