Anti-CASP3 p17 Rabbit Polyclonal Antibody [clone: D175]
Catalog # BWRLBS7073
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CASP3 p17
- Clonality:Polyclonal
- Clone:D175
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide conjugated to KLH derived from within residues 150 - 200 of Human active Caspase 3.
Caspase-3 p17 (D175) pAb detects endogenous levels of the large fragment (17 kDa) of activated caspase-3 resulting from cleavage adjacent to Asp175. This antibody does not recognize full length caspase-3 or other cleaved caspases.
Type: Primary
Antigen: CASP3 p17
Clonality: Polyclonal
Clone: D175
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat