Anti-p-FANCA Rabbit Polyclonal Antibody [clone: S1149]
BWRLBS4685
- Antibody type:Primary
- Antigen symbol:p-FANCA
- Clonality:Polyclonal
- Clone:S1149
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human FANCA around the phosphorylation site of Serine 1149.
p-FANCA (S1149) pAb detects endogenous levels of FANCA protein only when phosphorylated at Ser1149.
Type: Primary
Antigen: p-FANCA
Clonality: Polyclonal
Clone: S1149
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat