Anti-p-Smad1 Rabbit Polyclonal Antibody [clone: S465]
BWRLBS4573
- Antibody type:Primary
- Antigen symbol:p-Smad1
- Clonality:Polyclonal
- Clone:S465
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human Smad1 around the phosphorylation site of Serine 465.
p-Smad1 (S465) pAb detects endogenous levels of Smad1 protein only when phosphorylated at Ser465.
Type: Primary
Antigen: p-Smad1
Clonality: Polyclonal
Clone: S465
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat