Anti-p-VEGFR1/Flt-1 Rabbit Polyclonal Antibody [clone: Y1333]
BWRLBS4538
- Antibody type:Primary
- Antigen symbol:p-VEGFR1/Flt-1
- Clonality:Polyclonal
- Clone:Y1333
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human VEGFR1 around the phosphorylation site of Tyrosine 1333.
p-VEGFR1/Flt-1 (Y1333) pAb detects endogenous levels of VEGFR1/Flt-1 protein only when phosphorylated at Tyr1333
Type: Primary
Antigen: p-VEGFR1/Flt-1
Clonality: Polyclonal
Clone: Y1333
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat