Anti-p-MITF Rabbit Polyclonal Antibody [clone: S180]
BWRLBS4328
- Antibody type:Primary
- Antigen symbol:p-MITF
- Clonality:Polyclonal
- Clone:S180
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human MITF around the phosphorylation site of Serine 180.
p-MITF (S180) pAb detects endogenous levels of MITF protein only when phosphorylated at Ser180
Type: Primary
Antigen: p-MITF
Clonality: Polyclonal
Clone: S180
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat