- Antibody type:Primary
- Antigen symbol:p-NOS1
- Clonality:Polyclonal
- Clone:S852
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human NOS1 around the phosphorylation site of Serine 852.
p-NOS1 (S852) pAb detects endogenous levels of NOS1 protein only when phosphorylated at Ser852
Type: Primary
Antigen: p-NOS1
Clonality: Polyclonal
Clone: S852
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat