Anti-UCP3 Rabbit Polyclonal Antibody [clone: R308]
BWRLBS2849
- Antibody type:Primary
- Antigen symbol:UCP3
- Clonality:Polyclonal
- Clone:R308
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 280-320 of Human UCP3.
UCP3 (R308) pAb detects endogenous levels of UCP3 protein.
Type: Primary
Antigen: UCP3
Clonality: Polyclonal
Clone: R308
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat