Anti-PU.1 Rabbit Polyclonal Antibody [clone: R212]
Catalog # BWRLBS2649
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PU.1
- Clonality:Polyclonal
- Clone:R212
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 180-230 of Human PU.1.
PU.1 (R212) pAb detects endogenous levels of PU.1 protein.
Type: Primary
Antigen: PU.1
Clonality: Polyclonal
Clone: R212
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat