Anti-AIF-M1 Rabbit Polyclonal Antibody [clone: Y85]
BWRLBS2407
- Antibody type:Primary
- Antigen symbol:AIF-M1
- Clonality:Polyclonal
- Clone:Y85
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 61-110 of Human AIF-M1.
AIF-M1 (Y85) pAb detects endogenous levels of AIF-M1 protein.
Type: Primary
Antigen: AIF-M1
Clonality: Polyclonal
Clone: Y85
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat