Order Entry
Portugal
ContactUsLinkComponent
Anti-NMDAzeta1 Rabbit Polyclonal Antibody [clone: S885]
Anti-NMDAzeta1 Rabbit Polyclonal Antibody [clone: S885]
Catalog # BWRLBS1561
Anti-NMDAzeta1 Rabbit Polyclonal Antibody [clone: S885]
Catalog # BWRLBS1561

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    NMDA zeta1
  • Clonality:
    Polyclonal
  • Clone:
    S885
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide, corresponding to amino acids 860-910 of Human NMDAzeta1.

NMDAR1 (S885) pAb detects endogenous levels of NMDAR1 protein.

Type: Primary
Antigen: NMDAzeta1
Clonality: Polyclonal
Clone: S885
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat