Anti-ETS1 Rabbit Polyclonal Antibody [clone: A32]
Catalog # BWRLAP0387
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:ETS1
- Clonality:Polyclonal
- Clone:A32
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminus of Human ETS1.
ETS1 (A32) pAb detects endogenous levels of ETS1 protein.
Type: Primary
Antigen: ETS1
Clonality: Polyclonal
Clone: A32
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat