DNA purification kits, Maxwell® 16 cell LEV DNA purification kit
PROMAS1140
: Promega
:
- Pk:1 KIT
- Description:Maxwell® 16 Cell LEV DNA Purification Kit, 48 assays
- Sample size:48 preparations
The Maxwell® 16 Cell Low Elution Volume (LEV) DNA Purification Kit automatically extracts genomic DNA from sample types inherently low in cell number, such as amniotic fluid, cerebral spinal fluid (CSF) and cell culture supernatants. Purified genomic DNA can be easily obtained with samples containing less than 10000 cells. Used in combination with the Maxwell® 16 instrument.
- Process up to 16 samples in about 30 minutes
- High quality genomic DNA provides better performance in downstream applications
: The components in the kit can be ordered separately.
: The Maxwell® 16 instrument is supplied with pre-programmed purification procedures. The kit includes LEV plungers and LEV elution tubes.