PCR clean-up, Wizard® SV 96 PCR clean up system
PROMA9341
: Promega
:
- Pk:1
- Description:Wizard® SV 96 PCR Clean Up System
The Wizard® SV 96 PCR clean up system is designed for high throughput purification of 100 bp to 10 kb PCR products from excess nucleotides, primers and primer dimers. The purified DNA can be used for automated fluorescent sequencing, cloning, labeling, restriction digestion or microarray analysis without further manipulation. The Wizard® SV 96 PCR clean-up system uses 96-well filtration without the need to disassemble the manifold.
- Average recovery for 100 to 500 bp fragments of >90% and automated fluorescent sequencing Phred* 20 scores >600
- Membrane-based system allows recovery of >90% in as little as 20 minutes
- Filtrate waste is delivered directly to a vacuum trap
: The components in kit can be ordered separately.
: The complete kit includes membrane binding solution and Wizard® SV 96 binding plates.