Midipreps DNA purification system, Wizard® Plus
PROMA76400025
: Promega
:
- Pk:25
- Description:Wizard® Plus Midipreps DNA Purification System
The resin-based Wizard® Plus Midipreps DNA Purification System provides a simple and reliable method for rapid isolation of plasmid DNA. Midipreps may be processed individually or in multiples with the Vac-Man® (20 sample capacity) or Vac-Man® Jr. (2 sample capacity) laboratory vacuum manifold. The purified plasmid can be used directly for automated fluorescent DNA sequencing and restriction digestion without further manipulation and also can be used for in vitro transcription reactions.
- Purified DNA meets a target of >98% accuracy over 500 bases using pGEM®-3Zf(+) Vector in BigDye® terminator sequencing
- Entire procedure may be completed in 90 minutes or less
- Rapid batch column method used for DNA isolation
: The components in kit can be ordered separately.
: The complete kit includes Wizard® Midipreps DNA purification resin, Wizard® midicolumns, cell resuspension solution, cell lysis solution, neutralisation solution, column wash solution.