Order Entry
Portugal
ContactUsLinkComponent
RT-PCR system, GoTaq® 2-step system
RT-PCR system, GoTaq® 2-step system
Catalog # A6010
Supplier:  Promega
CAS Number:  
undefined
RT-PCR system, GoTaq® 2-step system
Catalog # A6010
Supplier:  Promega
CAS Number:  
Specifications
  • Pk:
    250 Tests
  • Description:
    GoTaq® 2-Step RT-PCR system, 5 ml
Specifications
About this item
The GoTaq® 2-Step RT-qPCR system facilitates detection and relative quantification of RNA expression levels via a two-step RT-qPCR method using integrated components such as GoScript™ reverse transcription system and GoTaq® qPCR master mix. It includes an optimised reaction buffer and M-MLV reverse transcriptase designed to enable efficient synthesis of first-strand cDNA in preparation for PCR amplification. The cDNA product can be added directly to downstream qPCR amplification reactions.
  • Sensitive detection for earlier quantitation of low and high copy number targets over a broad dynamic range
  • Resistant to a wide range of PCR inhibitors
  • Compatible with both fast and standard cycling methods on most Real-Time PCR instruments
  • Exceptional room temperature set-up is easy for automated and high throughput detection
Delivery: The complete system includes GoScript™ Reverse Transcriptase enzyme, GoScript™ reaction buffer(5X), GoTaq® qPCR master mix (2X), MgCl₂, PCR nucleotide mix, CXR reference dye, oligo(dT)15 primer, random primers, nuclease-free water and recombinant RNasin® ribonuclease inhibitor.