Plasmid DNA purification system, Wizard® SV 96
: Promega
The Wizard® SV 96 plasmid DNA purification systems provide a simple and reliable method for the rapid isolation of plasmid DNA using a silica membrane, 96-well, high throughput format. A single plate can be processed in 60 minutes or less. The purified plasmid can be used directly for automated fluorescent DNA sequencing as well as for other standard molecular biology techniques, including restriction enzyme digestion.
- Vac-Man® 96 vacuum manifold eliminates waste handling and allows simultaneous lysate clearing and DNA binding
- Novel plate design prevents cross-contamination during sample processing
- Flexibility – Designed for use in both manual and automated formats
- Purified DNA meets a target of >98% accuracy over 600 bases using pGEM®-3Zf(+) Vector DNA in BigDye® terminator sequencing
: The components in kit can be ordered separately.
: The complete kit includes Wizard® SV 96 neutralisation solution, Wizard® SV 96 lysate clearing plate, Wizard® SV 96 cell resuspension solution, cell lysis solution, 96-well deep well plates, binding plate, elution plate, plate sealers, column wash solution (CWA), alkaline protease solution and nuclease free water.
Frequently Bought Together