Genomic DNA purification system, Wizard® SV 96
Supplier: Promega
Certificates
About this item
The Wizard® SV 96 genomic DNA purification system provides a high-throughput, membrane based technique for consistent preparation of genomic DNA from cultured cells and tissue, including mouse tails. Amplifiable genomic DNA can be isolated from up to 5×10⁶ cells, 20 mg of tissue or up to 1,2 cm of a mouse tail tip without a centrifugation clearing step. With this system, genomic DNA is purified from cell lysates using 96-well vacuum filtration. Washing the bound DNA requires no disassembly of the manifold, and filtrate waste products are delivered directly to a vacuum trap, eliminating the need to empty waste collection trays.
- Genomic DNA can be obtained from mouse tails in 45 to 60 minutes and from cultured cells in 30 minutes
- No spins required
- High yield – purify 20 to 30 μg of DNA per prep from 1,2 cm of mouse tail
- Validated automated methods available
Ordering information: The components in the kit can be ordered separately.
Delivery: The complete kit includes 96-well deep well plates, binding plate, nuclei lysis solution, RNase A solution, column wash solution (CWA), nuclease free water, EDTA (0,5 M) pH 8,0, and Wizard® SV lysis buffer.
Frequently Bought Together