DNA purification systems, Wizard® Plus Maxipreps
Catalog # PROMA72700010
Supplier: Promega
CAS Number:
Specifications
- Pk:10
- Description:DNA purification system, Wizard® Plus Maxipreps
Specifications
About this item
The Wizard® Plus Maxipreps DNA purification system provides a simple and rapid resin based batch column method for purification of plasmid DNA that eliminates the need for cesium chloride:ethidium bromide gradient centrifugation. This system requires only a centrifuge, a vacuum source and a vacuum manifold. This system typically yields 300 μg to 1 mg of high copy number plasmid DNA (200 to 20000 bp) from a 100 to 500 ml culture in less than 3 hours. The purified DNA can be used directly for DNA sequencing and restriction digestion without further manipulation and also can be used in vitro transcription reactions.
- Purified DNA is suitable for restriction enzyme digestions, automated fluorescent DNA sequencing, transformation and subcloning
- Yields plasmid DNA of comparable quantity and quality, and eliminates the time and labour needed for caesium chloride:ethidium bromide gradient techniques
- Rapid batch binding and column washing method used for DNA isolation
Ordering information: The components in kit can be ordered separately.
Delivery: The complete kit includes Wizard® Maxipreps DNA purification resin, cell resuspension solution, cell lysis solution, neutralisation solution, Maxi/Megapreps filtering system and column wash solution.