Order Entry
Portugal
ContactUsLinkComponent
 
DNA purification systems, Wizard® Plus SV minipreps
 :  Promega
 
undefined
DNA purification systems, Wizard® Plus SV minipreps
 :  Promega

 

  • Application:
  • Method / Format:
  • Refine target molecule:
  • Sample type:
  • Target molecule:
  • Environmentally Preferable:

 

Frequently Bought Together