Dextran sulphate sodium salt, M.W. ∼36000 - 50000, white powder, Colitis Grade
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:9011-18-1
:
Formula:(C₆H₇Na₃O₁₄S₃)n
MDL Number:MFCD00081551
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Moisture:<10%
- Residue on Ignition:<50%
- Sulphur :~18 - 20%
- Glucose:35 - 50%
- pH (1% aq soln):5 - 7,5
- Specific Rotation :+75° to +105°
- Free Sulphate:≤0,2%
Specification Test Results
Frequently Bought Together