Human recombinant ATG5 (from E. coli)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:APG5 although stable at 4 °C for week, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA).Please prevent freeze-thaw cycles.
- Protein synonyms:Autophagy
- Protein/peptide name:ATG5
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Formulation:APG5 containing 10mM Tris, pH 8.0, 0.01% Triton X-100 and 0.002% NaN3 and 10mM DTT.
- Tested applications:SDS-PAGE
Frequently Bought Together